Organic reference standard, Dimethylnitrosoamine 10 µg/ml in methanol, Dr Ehrenstorfer
EHERL15604000ME
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:62-75-9
- Description:Dimethylnitrosoamine 10 µg/ml in methanol
- Formula:C₂H₆N₂O
- Description:Dimethylnitrosoamine