0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Netherlands
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standard, p-Cresol 10 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, p-Cresol 10 µg/ml in methanol, Dr Ehrenstorfer
EHERL15140400ME
:
EHRENSTORFER
:
Organic reference standard, p-Cresol 10 µg/ml in methanol, Dr Ehrenstorfer
EHERL15140400ME
:
EHRENSTORFER
:
Pack type:
Glass bottle
Pk:
10 mL
CAS No.:
106-44-5
Description:
p-Cresol 10 µg/ml in methanol
Formula:
C₇H₈O
Description:
p-Cresol