Organic reference standard, Fluorobenzene 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL13781000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:462-06-6
- Description:Fluorobenzene 10 µg/ml in methanol
- Formula:C₆H₅F
- Description:Fluorobenzene