Order Entry
Netherlands
ContactUsLinkComponent
2,6-Toluenediisocyanate
2,6-Toluenediisocyanate
Catalog # EHERCA17594600
Supplier:  EHRENSTORFER
CAS Number:  
2,6-Toluenediisocyanate
Catalog # EHERCA17594600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :174.16
EINECS:202-039-0
UN:2078
ADR:6.1,II
CAS Number:91-08-7
Formula:C₉H₆N₂O₂
MDL Number:MFCD00002010