Order Entry
Netherlands
ContactUsLinkComponent
Methyl-4-fluoro-2-methoxybenzoate ≥97%
Methyl-4-fluoro-2-methoxybenzoate ≥97%
Catalog # H52370.06
CAS Number:  
Methyl-4-fluoro-2-methoxybenzoate ≥97%
Catalog # H52370.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:204707-42-6
 :
Formula:C₉H₉FO₃
 :184.17 g/mol
Storage Temperature:Ambient
MDL Number:MFCD06203990