

2,4-Difluoro-3-methoxyaniline ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:886499-08-7
:
Formula:C₇H₇F₂NO
MDL Number:MFCD04115929
:159.14 g/mol
UN:2810
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together