



2-Ethylphenylhydrazine hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19398-06-2
:58711-02-7
Formula:C₈H₁₃ClN₂
MDL Number:MFCD00071599
:172.66 g/mol
Melting Pt:∼ 178 °C (dec.)
Density:1.21
EINECS:421-460-2
ADR:9,III
:GHS06
:GHS08
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together