Methyl-5-chloro-2-methoxybenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:33924-48-0
:
Formula:C₉H₉ClO₃
MDL Number:MFCD00017511
:200.62 g/mol
Boiling Pt:235…240 °C
Density:1.259
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:251-743-4
Related Information
Frequently Bought Together