
Thiocarbonyl chloride 85%, AcroSeal®
Catalog # 426441000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Orange-red to red Liquid
- GC:≥80 %
- Refractive index:1.4025 to 1.4065 (20°C, 589 nm)
Specification Test Results
About this item
:
:114.98 g/mol
Flash Pt:62 °C
Storage Temperature:Refrigerator
EINECS:207-341-6
UN:2474
ADR:6.1,I
:GHS06
CAS Number:463-71-8
Formula:CCl₂S
MDL Number:MFCD00004918