Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
2,2,2-Trifluoroethyl methacrylate 99% stabilised
  ACRO312780250
 :  
undefined
 
 
2,2,2-Trifluoroethyl methacrylate 99% stabilised
  ACRO312780250
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3600 to 1.3620 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Stabilizer:
    100 ppm MEHQ

 

 

 :
 :168.12 g/mol
 :59 °C (100 mmHg)
 :16 °C
 :Refrigerator
 :206-525-3
 :1993
 :3,II
 :GHS07
 :GHS02
 :352-87-4
 :C₆H₇F₃O₂
 :MFCD00013576