Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
p-Toluenesulphonyl isocyanate 96%
 
undefined
 
 
p-Toluenesulphonyl isocyanate 96%

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5330 to 1.5360 (20°C, 589 nm)
  • GC:
    ≥95 %

 

 

Frequently Bought Together