
5-Bromo-2-chlorobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21739-92-4
:
Formula:C₇H₄BrClO₂
MDL Number:MFCD00002415
:235.46 g/mol
Boiling Pt:316 °C (1013 hPa)
Melting Pt:157…162 °C
Storage Temperature:Ambient
EINECS:244-558-5
:GHS07
Related Information
Frequently Bought Together