Order Entry
Netherlands
ContactUsLinkComponent
1-Octadecene
1-Octadecene
Catalog # EHERC15710400
Supplier:  EHRENSTORFER
CAS Number:  
1-Octadecene
Catalog # EHERC15710400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :252.48 g/mol
Boiling Pt:315 °C (1013 hPa)
Boiling Pt:315 °C (1013 hPa)
Melting Pt:17.5 °C
Density:0.789 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-012-9
CAS Number:112-88-9
Formula:C₁₈H₃₆
MDL Number:MFCD00009003