Order Entry
Netherlands
ContactUsLinkComponent
 
 
1-Octadecene 90% tech.
 
undefined
 
1-Octadecene 90% tech.

 

  • Appearance:
    Clear colorless to amber Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4435 to 1.4455 (20°C, 589 nm)
  • GC:
    ≥88 %

 

 

Frequently Bought Together