Order Entry
Netherlands
ContactUsLinkComponent
 
Health
Irritant
Trichloroethylene
Catalog # PROOCHE05
CAS Number:  
undefined
Health
Irritant
Trichloroethylene
Catalog # PROOCHE05
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,5 mL
  • Seq:
    2095

Specifications

About this item

 :
Merck Index:12,09769
Boiling Pt:86.7 °C (1013 hPa)
Melting Pt:–73 °C
Density:1.46 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-167-4
UN:1710
ADR:6.1,III
 :131.39 g/mol
 :GHS08
 :GHS07
CAS Number:79-01-6
Formula:ClCH=CCl₂
MDL Number:MFCD00000838