Order Entry
Netherlands
ContactUsLinkComponent
Tetrachloroethylene
  PROOHDF12
 :  
undefined
Tetrachloroethylene
  PROOHDF12
 :  
  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275
 :
 :165.83 g/mol
 :12,09332
 :121.1 °C (1013 hPa)
 :–22 °C
 :1.63 g/cm³ (20 °C)
 :Ambient
 :204-825-9
 :1897
 :6.1,III
 :GHS08
 :GHS09
 :127-18-4
 :C₂Cl₄
 :MFCD00000834