



2-Methylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-83-5
:
Formula:C₆H₁₄
MDL Number:MFCD00009406
Boiling Pt:60 °C (760 mmHg)
Melting Pt:–154 °C
Density:0.66 g/cm³
Flash Pt:–23 °C
Storage Temperature:Ambient
EINECS:203-523-4
ADR:3,II
:86.18 g/mol
UN:1208
:GHS07
:GHS08
:GHS09
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3700 to 1.3720 (20°C, 589 nm)
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together