2,2,4-Trimethylpentane ≥99.8% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # 9335-22
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 5
- FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤10
- ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL):≤5
- Assay ((CH₃)₃CCH₂CH(CH₃)₂) (by GC, corrected for water):≥99.8 %
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Water (by KF, coulometric):≤0.03 %
Specification Test Results
About this item
Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
:
:114.23 g/mol
EINECS:208-759-1
UN:1262
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:540-84-1
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
Boiling Pt:99.2
Melting Pt:–107 °C
Density:0.692 g/cm³ (20 °C)
Flash Pt:–12 °C
ADR:3 II
Merck Index:13,05212
:Packaged under inert gas.