tert-Butylamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-64-9
:
Formula:C₄H₁₁N
MDL Number:MFCD00008050
:73.14 g/mol
Merck Index:14,1545
Boiling Pt:44…46 °C
Melting Pt:–67 °C
Density:0.696
Flash Pt:–38 °C (–36 °F)
Storage Temperature:Ambient
EINECS:200-888-1
UN:2733
ADR:3,II
:GHS02
:GHS06
:GHS05
Related Information
Frequently Bought Together