2,2,6,6-Tetramethyl-3,5-heptanedione ≥98%
A15118.22
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
:
:184.28 g/mol
:72…73 °C (6 mmHg)
:28…30 °C
:0.889
:67 °C (152 °F)
:Ambient
:214-268-3
:GHS07
:1118-71-4
:C₁₁H₂₀O₂
:MFCD00008848