3-Methylbutanone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Isopropyl Methyl Ketone, 3-Methyl-2-butanone
CAS Number:563-80-4
:
Formula:C₅H₁₀O
MDL Number:MFCD00008919
Boiling Pt:94
Melting Pt:–92 °C
Density:0.8 g/cm³
Molecular Weight:86.13 g/mol
Flash Pt:6 °C
UN:2397
ADR:3 II
Merck Index:12,06164
EINECS:209-264-3
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3855 to 1.3905 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together