n-Pentane
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Pentane
- CAS Number:109-66-0
- Formula:H₃C(CH₂)₃CH₃
- MDL Number:MFCD00009498
- Molecular Weight:72.15 g/mol
- Merck Index:13,07193
- Boiling Pt:35…36 °C (1013 hPa)
- Melting Pt:–130 °C
- Density:0.63 g/cm³ (20 °C)
- Flash Pt:–40 °C
- Storage Temperature:Ambient
- EINECS:203-692-4
- UN:1265
- ADR:3,II
- REACH:01-2119459286-30
2 Options Available Below
Product Details & Documents
Synonyms: Pentane
- CAS Number:109-66-0
- Formula:H₃C(CH₂)₃CH₃
- MDL Number:MFCD00009498
- Molecular Weight:72.15 g/mol
- Merck Index:13,07193
- Boiling Pt:35…36 °C (1013 hPa)
- Melting Pt:–130 °C
- Density:0.63 g/cm³ (20 °C)
- Flash Pt:–40 °C
- Storage Temperature:Ambient
- EINECS:203-692-4
- UN:1265
- ADR:3,II
- REACH:01-2119459286-30
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



