Order Entry
Netherlands
ContactUsLinkComponent
n-Pentane 99+% for HPLC
Irritant
Health
Environment
Flammable
n-Pentane 99+% for HPLC
Certificates
undefined
Irritant
Health
Environment
Flammable
n-Pentane 99+% for HPLC

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.01 %
  • GC:
    ≥99.0 %
  • UV:
    at 230 nm A: ≤0.03
  • Non-volatile matter:
    ≤0.0003 %
  • Free acid (CH3COOH):
    ≤0.0005 %

Specification Test Results

Related Information

Frequently Bought Together