

cis-1,3-Pentadiene ≥97.0% (by GC) stabilised
: TCI
:1574-41-0
:
:C₅H₈
:MFCD00063977
:68.12 g/mol
:43°C
:–140°C
:216-401-0
:3295
:3,II
:Stabilised with 4-tert-butylpyrocatechol (TBC)
:GHS02
:GHS07
:Freezer
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together