Order Entry
Netherlands
ContactUsLinkComponent
1H,1H,9H-Hexadecafluoro-1-nonanol ≥97%
1H,1H,9H-Hexadecafluoro-1-nonanol ≥97%
Catalog # B21407.22
CAS Number:  
1H,1H,9H-Hexadecafluoro-1-nonanol ≥97%
Catalog # B21407.22
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

CAS Number:376-18-1
 :
Formula:C₉H₄F₁₆O
MDL Number:MFCD00039629
 :432.1 g/mol
Boiling Pt:157…158 °C (200 mmHg)
Melting Pt:55…60 °C
Storage Temperature:Ambient
EINECS:206-806-0