Order Entry
Netherlands
ContactUsLinkComponent
Diatomaceous earth filter papers, grade 470
 :  Sartorius
undefined
Diatomaceous earth filter papers, grade 470
 :  Sartorius
  • Catalog No:
  • SARTFT-4-606-320
  • SARTFT-4-606-240
  • SARTFT-4-606-185
  • SARTFT-4-606-150
  • SARTFT-4-606-125
  • Weight:
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • Thickness:
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • 0,32 mm
  • Filtration speed:
  • 80 s
  • 80 s
  • 80 s
  • 80 s
  • 80 s
Frequently Bought Together