n-Dodecane
Glass bottle, 1
Catalog # METT51338012
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
Synonyms: Dodecane
- CAS Number:112-40-3
- Formula:H₃C(CH₂)₁₀CH₃
- MDL Number:MFCD00008969
- Molecular Weight:170.34 g/mol
- Boiling Pt:216 °C (1013 hPa)
- Melting Pt:–9.6 °C
- Density:0.7487 g/cm³ (20 °C)
- Flash Pt:74 °C
- Storage Temperature:Ambient
- EINECS:203-967-9
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1



