Order Entry
Netherlands
Orders LinkContactUsLinkComponent
1,5-Pentanedithiol 96%

1,5-Pentanedithiol 96%

Irriterend
Leverancier:   Thermo Fisher Scientific
1,5-Pentanedithiol 96%
Certificates
1,5-Pentanedithiol 96%
1,5-Pentanedithiol 96%

2 Opties Hieronder Beschikbaar

Product Details & Documents

Purity: 96% CAS No.: 928-98-3 Molecular Formula: HS(CH2)5SH Formula Weight: 136.28 Flash Point: 95°(203°F) Refractive Index: 1.5194 Odour: Unpleasant Melting Point: -72° Boiling Point: 216-218° Beilstein Reference: 1732335 UN No.: 3334 MDL No.: MFCD00004908

Synoniemen:  1,5-Dimercaptopentane
  • CAS-nummer:928-98-3
  • Formule:C₅H₁₂S₂
  • MDL-nummer:MFCD00004908
  • Molecular Weight:136.28 g/mol
  • Kokend Pt:216…218 °C
  • Smelten Pt:–72 °C
  • Dichtheid:1.016
  • Flits Pt:95 °C (203 °F)
  • Temperatuur bij opslag:Ambient
  • EINECS:213-194-9
  • ADR:9,III
Product Family Options

Productinformatie

Aanbevelingen

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.