

1-Hexanethiol 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-31-9
:
Formula:C₆H₁₄S
MDL Number:MFCD00004909
Boiling Pt:150…154 °C (760 mmHg)
Melting Pt:81…80 °C
Density:0.84 g/cm³
Flash Pt:30 °C
Storage Temperature:Ambient
EINECS:203-857-0
ADR:6.1,II
:118.24 g/mol
UN:3071
:GHS06
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4480 to 1.4520 (20°C, 589 nm)
- Density:(20°C) 0.836 to 0.844 g/ml
- GC:≥95 %
Specification Test Results
Frequently Bought Together