Bestelling invoeren
Netherlands
ContactUsLinkComponent
Ethanol for molecular biology, Sigma-Aldrich®
Flammable
Irritant
Ethanol for molecular biology, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
Ethanol for molecular biology, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Appearance (colour):
    colourless
  • Appearance (description):
    clear
  • Purity (GC):
    ≥ 99.8 %
  • Acidity:
    ≤ 0.0002 meq/g
  • Alkalinity:
    ≤ 0.0002 meq/g
  • Heavy metals (as Pb):
    ≤ 0.0001 %
  • DNases (nicking-activity):
    not detectable
  • RNases:
    not detectable

Specification Test Results

Related Information

Frequently Bought Together