1-Heptanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Heptyl alcohol
CAS Number:111-70-6
:
Formula:H₃C(CH₂)₅CH₂OH
MDL Number:MFCD00002986
:116.2 g/mol
Merck Index:13,04681
Boiling Pt:176 °C (1013 hPa)
Melting Pt:–35 °C
Density:0.82 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:70 °C
EINECS:203-897-9
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:1 g/l (18 °C)
- Melting Point:-35 °C
- Molar Mass:116.2 g/mol
- Boiling Point:176 °C (1013 hPa)
- Vapor Pressure:0.5 hPa (20 °C)
- Flash Point:78 °C
- Refractive Index:1.4242 (20 °C, 589 nm)
- Explosion Limit:0.9 %(V)
- Density:0.82 g/cm3 (20 °C)
- Ignition Point:350 °C
Specification Test Results
Related Information
Frequently Bought Together