Order Entry
Netherlands
ContactUsLinkComponent
1-Butanol
Flammable
Corrosive
Irritant
1-Butanol
Catalog # ROTHT178.1
Supplier:  Roth Carl
CAS Number:  
undefined
Flammable
Corrosive
Irritant
1-Butanol
Catalog # ROTHT178.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

About this item

 :
 :74.12 g/mol
Merck Index:13,01539
Boiling Pt:117.6 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.812 g/cm³ (20 °C)
Flash Pt:30 °C
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
REACH:01-2119484630-38
 :GHS02
 :GHS05
 :GHS07
CAS Number:71-36-3
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964