


Tributylamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:102-82-9
:
Formula:C₁₂H₂₇N
MDL Number:MFCD00009431
:185.35 g/mol
Merck Index:13,09691
Boiling Pt:216 °C (1013 hPa)
Melting Pt:–70 °C
Density:0.7771 g/cm³ (20 °C)
EINECS:203-058-7
UN:2542
ADR:6.1,II
:M-Clarity™ quality level = MQ200
Flash Pt:63 °C
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.3 g/l (20 °C)
- Melting Point:-70 °C
- Molar Mass:185.35 g/mol
- Boiling Point:216 - 217 °C (1013 hPa)
- Vapor Pressure:0.18 hPa (20 °C)
- Flash Point:63 - 66 °C
- Explosion Limit:1.4 - 6.0 %(V)
- Density:0.78 g/cm3 (20 °C)
- pH:10.6 (H2O, 20 °C) (saturated aqueous solution)
- Ignition Point:190 °C
Specification Test Results
Frequently Bought Together