

2,2,3,3-Tetrafluoro-1-propanol 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-37-9
:
Formula:C₃H₄F₄O
MDL Number:MFCD00004676
:132.06 g/mol
Merck Index:12,09346
Boiling Pt:109 °C (760 mmHg)
Melting Pt:–15 °C
Density:1.47 g/cm³
Flash Pt:43 °C
Storage Temperature:Ambient
EINECS:200-955-5
UN:1986
ADR:3,III
:GHS02
:GHS06
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3200 to 1.3220 (20°C, 589 nm)
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together