Order Entry
Netherlands
ContactUsLinkComponent
4-Chloro-1-butanol 85% balance THF and HCl
Flammable
Corrosive
Irritant
Health
4-Chloro-1-butanol 85% balance THF and HCl
Certificates
undefined
Flammable
Corrosive
Irritant
Health
4-Chloro-1-butanol 85% balance THF and HCl

Specification Test Results

  • Appearance:
    Clear colorless to light yellow to light pink Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4510 to 1.4630 (20°C, 589 nm)
  • GC:
    ≥80 %

Specification Test Results

Related Information

Frequently Bought Together