


Dichloromethyl methyl ether for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4885-02-3
:
Formula:C₂H₄Cl₂O
MDL Number:MFCD00000828
:114.96 g/mol
Density:1.29 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:225-498-9
UN:2924
ADR:3 (8),II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS05
:GHS07
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Molar Mass:114.95 g/mol
- Boiling Point:85 - 87 °C (1013 hPa)
- Vapor Pressure:84 hPa (20 °C)
- Flash Point:42 °C
- Refractive Index:1.43 (20 °C, 589 nm)
- Density:1.29 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together