


Trichloroacetic acid (TCA) glacial Ph. Eur.
:76-03-9
:
:Cl₃CCOOH
:MFCD00004177
:163.39 g/mol
:13,09700
:196 °C (1013 hPa)
:54…56 °C
:1.63 g/cm³ (20 °C)
:113 °C (closed cup)
:Ambient
:200-927-2
:1839
:8,II
:GHS05
:GHS07
:GHS09
- Assay:98.0 - 100.5 %
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Appearance of solution:Passes test
- Cl (Chloride):Max. 100 ppm
- Sulphated ash:Max. 0.1 %
- Residual solvents:Passes test
Frequently Bought Together