Order Entry
Netherlands
ContactUsLinkComponent
1,3-Dichloropropane
1,3-Dichloropropane
Catalog # EHERCA12481000
Supplier:  EHRENSTORFER
CAS Number:  
1,3-Dichloropropane
Catalog # EHERCA12481000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :112.99 g/mol
Boiling Pt:120.4 °C (1013 hPa)
Boiling Pt:120,4 °C (1013 hPa)
Melting Pt:–99.5 °C
Melting Pt:–99,5 °C
Density:1,1717 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-531-3
UN:1993
ADR:3,II
CAS Number:142-28-9
Formula:ClH₂CCH₂CH₂Cl
MDL Number:MFCD00000999