Order Entry
Netherlands
ContactUsLinkComponent
Sodium propionate 99.0-100.5%
Irritant
Sodium propionate 99.0-100.5%
Certificates
undefined
Irritant
Sodium propionate 99.0-100.5%
Specification Test Results
  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤1 % (K.F.)
  • Titration with HClO4:
    99.0 to 100.5 % (On dried substance)
  • Iron (Fe):
    ≤30 ppm
  • Heavy metals (as Pb):
    ≤10 ppm