Bestelling invoeren
Netherlands
Orders LinkContactUsLinkComponent
Ethylacetaat

Ethylacetaat

Glass bottle, 2,5 L
Catalog # MSPPG2015
Flammable
Irritant
Marketsource
   Veiligere gezondheid van mens en milieu
Ethylacetaat
Catalog # MSPPG2015
Marketsource
   Veiligere gezondheid van mens en milieu
CAS Number:  141-78-6
Ethylacetaat
Ethylacetaat

Glass bottle, 2,5 L

CAS Number:  141-78-6
Price
Inquire For Price
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Acetic acid ethyl ester
  • CAS Number:141-78-6
  • Formula:CH₃COOC₂H₅
  • MDL Number:MFCD00009171
  • Molecular Weight:88.11 g/mol
  • Merck Index:13,03792
  • Boiling Pt:77,1 °C (1013 hPa)
  • Melting Pt:–83 °C
  • Density:0,902 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Kamertemperatuur
  • EINECS:205-500-4
  • UN:1173
  • ADR:3,II
  • REACH:01-2119475103-46
Specifications

Specifications

  • Verpakking:
    Glass bottle
  • VPE:
    2,5 L
  • Cat. No.:
    MSPPG2015
  • Milieu certificering:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Veiligere gezondheid van mens en milieu

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.