Methyl ethyl ketone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:78-93-3
:
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648
Merck Index:13,06097
Boiling Pt:79.6 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.8054 g/cm³ (20 °C)
Flash Pt:–4 °C (DIN 51755 Part 1)
Storage Temperature:Ambient
UN:1193
ADR:3,II
:72.11 g/mol
EINECS:201-159-0
REACH:01-2119457290-43
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay:Min. 99 %
- Boiling point:79 - 80 °C
- Density (20/4):0.804 - 0.807
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC25654.292 spec_VWRC25654.320 spec_VWRC25654.361 spec_VWRC25654.361
Product specification
VWR® Green Solvents for Sustainable Chemistry.pdf
Product Literature
Frequently Bought Together