Methyl ethyl ketone 99+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-93-3
:
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648
Merck Index:14,6072
Boiling Pt:80 °C
Melting Pt:–87 °C
Density:0.805
Flash Pt:–3 °C (26 °F)
Storage Temperature:Ambient
UN:1193
ADR:3,II
:72.11 g/mol
EINECS:201-159-0
REACH:01-2119457290-43
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Frequently Bought Together