Methyl ethyl ketone ACS
:78-93-3
:
:C₂H₅COCH₃
:MFCD00011648
:13,06097
:80 °C (760 mmHg)
:–87 °C
:0.806 g/cm³
:–7 °C
:Ambient
:1193
:3,II
:72.11 g/mol
:201-159-0
:01-2119457290-43
:GHS07
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Appearance:Clear liquid
- Water:≤0.20 %
- GC:≥99.0 %
- Color scale:≤15 APHA
- Residue after evaporation:≤0.0025 %
- Titrable acid:≤0.0005 meq/g
Frequently Bought Together