
Triethylammonium chloride ≥99.0% for HPLC, Fluka™
96249-250G
:
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
- Identity (NMR):complying
- Assay:min. 99.0 %
- Colour of the solution:complying
- Clearness of the solution:complying
- Absorbance 500 nm:max. 0.02
- Absorbance at 260 nm:max. 0.02
- Absorbance at 250 nm:max. 0.03
- Absorbance at 230 nm:max. 0.04
- Absorbance at 220 nm:max. 0.05
- Suitability test (HPLC):complying
:
:137.65 g/mol
:0,7637 g/cm³ (20 °C)
:209-067-2
:GHS07
:Room Temperature
:554-68-7
:C₆H₁₆ClN
:MFCD00012500