Digitoxin
Glass bottle for solids, 50 mg
Catalog # ROTH7359.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:71-63-6
- Formula:C₄₁H₆₄O₁₃
- MDL Number:MFCD00003686
- Molecular Weight:764.95 g/mol
- Melting Pt:276 °C
- EINECS:200-760-5
- UN:2811
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Cat. No.:ROTH7359.1



