Organic reference standard, Coronene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20675000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:191-07-1
- Description:Coronene 10 µg/ml in cyclohexane
- Formula:C₂₄H₁₂
- Description:Coronene