Order Entry
Netherlands
ContactUsLinkComponent
Benzamidine hydrochloride hydrate 98%
Irritant
Benzamidine hydrochloride hydrate 98%
Certificates
undefined
Irritant
Benzamidine hydrochloride hydrate 98%

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    97.5 to 102.5 % (on dry substance)
  • Water:
    ≤19 % (K.F.)
  • Molar absorption coefficient:
    ≥45000 l/(mol.cm) (at Lambda max)

Specification Test Results

Related Information

Frequently Bought Together