Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Dexamethasone

Glass bottle for solids, 100 mg
Catalog # PROOICRS0267
Dexamethasone
Catalog # PROOICRS0267
CAS Number:  50-02-2
Dexamethasone

Glass bottle for solids, 100 mg

CAS Number:  50-02-2
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
  • CAS Number:50-02-2
  • Formula:C₂₂H₂₉FO₅
  • MDL Number:MFCD00064136
  • Molecular Weight:392.47 g/mol
  • Melting Pt:268…271 °C
  • EINECS:200-003-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications