Dexamethasone
Glass bottle for solids, 100 mg
Catalog # SANTSC-29059
Marketsource
Supplier: Santa Cruz Biotechnology
Dexamethasone
Catalog # SANTSC-29059
Supplier: Santa Cruz Biotechnology
Marketsource
CAS Number: 50-02-2
Jump to:
Product Details & Documents
Synonyms: Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
- CAS Number:50-02-2
- Formula:C₂₂H₂₉FO₅
- MDL Number:MFCD00064136
- Molecular Weight:392.47 g/mol
- Melting Pt:268…271 °C
- EINECS:200-003-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



