4-Bromo-2,5-difluorobenzenesulphonyl chloride
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:207974-14-9
:
Formula:C₆H₂BrClF₂O₂S
MDL Number:MFCD00042181
:291.5 g/mol
Boiling Pt:76 °C (0.01 mmHg)
Melting Pt:34…36 °C
UN:3261
ADR:8,II
Frequently Bought Together